Merck
Sublytic C5b-9 Induces Glomerular Mesangial Cell Apoptosis Through miR-3546/SOX4/Survivin Axis in Rat Thy-1 Nephritis.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-09-21)
Chunyan Yao, Fengxia He, Longfei Liu, Zhiwei Zhang, Chenhui Zhao, Wen Qiu, Dan Zhao, Jing Zhang, Mengxiao Xie, Yajuan Gong, Tianyi Yu, Lu Xia, Baomei Qian, Yingwei Wang
ABSTRAKT
The activation of complement system and the formation of C5b-9 complex have been confirmed in the glomeruli of patients with mesangioproliferative glomerulonephritis (MsPGN). However, the role and mechanism of C5b-9-induced injury in glomerular mesangial cell (GMC) are poorly understood. Rat Thy-1N is an animal model for studying MsPGN. It has been revealed that the attack of C5b-9 to the GMC in rat Thy-1N is sublytic, and sublytic C5b-9 can cause GMC apoptosis, but the underlying mechanism is not fully elucidated. To explore the role and regulatory mechanism of C5b-9 in MsPGN lesion, we used rat Thy-1N model and first detected the change of microRNA (miRNA) profiles both in Thy-1N rat renal tissues (in vivo) and in the cultured GMCs with sublytic C5b-9 stimulation (in vitro). Then we determined the effect of miR-3546, which increased both in vivo and in vitro, on GMC apoptosis upon sublytic C5b-9 as well as the involved mechanism. Rat Thy-1N model was established and GMCs were treated with sublytic C5b-9. The rat renal cortex and the stimulated GMCs were obtained for miRNA microarray detection. Subsequently, the increased miRNAs were verified by real-time PCR. Meanwhile, to ascertain the ability of some miRNAs to upregulate cleaved caspase 3 and induce GMC apoptosis, the corresponding miRNA mimics were transfected into GMCs, followed by western blotting (WB) and flow cytometry mesurement. Thereafter, the miR-3546-targeted gene (SOX4) was predicted using bioinformatics approaches, and SOX4 expression in Thy-1N tissues and in the GMCs upon sublytic C5b-9 stimulation or miR-3546 mimic/inhibitor transfection were detected using real-time PCR and WB. To prove that miR-3546 can affect SOX4 gene transcription and SOX4 can regulate survivin expression, dual luciferase reporter assay, real-time PCR, WB and chromatin immunoprecipitation (ChIP) assays were performed. Furthermore, the role of miR-3546/SOX4/survivin axis in the GMC apoptosis induced by sublytic C5b-9 was examined using WB and flow cytometry. Compared with normal renal tissues and untreated GMCs, there were 43 and 62 upregulated miRNAs (> 2-fold) in Thy-1N tissues and sublytic C5b-9-stimulated GMCs respectively. A total of 17 miRNAs were increased both in vivo and in vitro, 11 of which were validated by real-time PCR. Among them, miR-3546 could markedly promote GMC apoptosis and inhibit SOX4 or survivin expression in response to sublytic C5b-9, and either SOX4 or survivin overexpression markedly rescued the GMC apoptosis mediated by miR-3546 mimic. Additionally, SOX4 overexpression could reverse the survivin suppression by miR-3546 mimic, and SOX4 could bind to survivin promoter (-1,278 to -853 nt) and activate survivin gene transcription. MiR-3546/ SOX4/survivin axis has a promoting role in the GMC apoptosis triggered by sublytic C5b-9, and our findings may provide a new insight into the pathogenesis of rat Thy-1N and human MsPGN.