Merck
  • Inhibitory Effect of PgAFP and Protective Cultures on Aspergillus parasiticus Growth and Aflatoxins Production on Dry-Fermented Sausage and Cheese.
Inhibitory Effect of PgAFP and Protective Cultures on Aspergillus parasiticus Growth and Aflatoxins Production on Dry-Fermented Sausage and Cheese.
Microorganisms (2018-07-18)
Josué Delgado, Alicia Rodríguez, Alfredo García, Félix Núñez, Miguel A Asensio
ABSTRAKT
Aflatoxigenic molds can grow and produce aflatoxins on dry-fermented meat and cheese. The small, basic, cysteine-rich antifungal protein PgAFP displays a time-limited inhibitory ability against unwanted molds by increasing reactive oxygen species (ROS), which can lead to increased aflatoxin production. However, calcium abolishes the inhibitory effect of PgAFP on certain Aspergillus spp. To maximize the antifungal effect, this protein may be combined with protective cultures. Yeasts and lactic acid bacteria may counteract the impact of calcium on PgAFP fungal inhibition. The objective of this work was to study the effect of PgAFP and different combined treatments with Debaryomyces hansenii and/or Pediococcus acidilactici against growth of and aflatoxin production by an aflatoxigenic strain of Aspergillus parasiticus in both culture media and dry-fermented foods with low or high calcium levels. Aflatoxins production was increased by PgAFP but dramatically reduced by P. acidilactici in low calcium culture medium, whereas in the Ca-enriched culture medium, all treatments tested led to low aflatoxins levels. To study whether PgAFP and the protective microorganisms interfere with ROS and aflatoxin production, the relative expression of genes foxA, which is involved in peroxisomal β-oxidation, and aflP, which is required for aflatoxin biosynthesis, were evaluated. The aflatoxin overproduction induced by PgAFP seems not to be linked to peroxisomal β-oxidation. The combination of PgAFP and D. hansenii provided a successful inhibitory effect on A. parasiticus growth as well as on aflatoxin production on sliced dry-fermented sausage and cheese ripened up to 15 days, whereas P. acidilactici did not further enhance the protective effect of the two former agents. Therefore, the combined treatment of PgAFP and D. hansenii seems to provide a promising protective mean against aflatoxin-producing A. parasiticus on dry-fermented foods.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Phenyl-1-naphthylamine, reagent grade, 98%