Merck
Systematic Comparison of Strategies to Achieve Soluble Expression of Plasmodium falciparum Recombinant Proteins in E. coli.
Molecular biotechnology (2018-09-28)
Liliana Morales, Paula Hernández, Jacqueline Chaparro-Olaya
ABSTRAKT
Constructs containing partial coding sequences of myosin A, myosin B, and glideosome-associated protein (50 kDa) of Plasmodium falciparum were used to challenge several strategies designed in order to improve the production and solubility of recombinant proteins in Escherichia coli. Assays were carried out inducing expression in a late log phase culture, optimizing the inductor concentration, reducing the growth temperature for induced cultures, and supplementing additives in the lysis buffer. In addition, recombinant proteins were expressed as fusion proteins with three different tags (6His, GST, and MBP) in four different E. coli strains. We found that the only condition that consistently produced soluble proteins was the use of MBP as a fusion tag, which became a valuable tool for detecting the proteins used in this study and did not caused any interference in protein-protein interaction assays (Far Western Blot). Besides, we found that BL21-pG-KJE8 strain did not improve the solubility of any of the recombinant protein produced, while the BL21-CodonPlus(DE3)-RIL strain improved the expression of some of them independent of the rare codon content. Proteins with rare codons occurring at high frequencies (» 10%) were expressed efficiently in strains that do not supplement tRNAs for these triplets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology