Merck
  • Oro-gastro-intestinal digestion of starch in white bread, wheat-based and gluten-free pasta: Unveiling the contribution of human salivary α-amylase.
Oro-gastro-intestinal digestion of starch in white bread, wheat-based and gluten-free pasta: Unveiling the contribution of human salivary α-amylase.
Food chemistry (2018-10-31)
Daniela Freitas, Steven Le Feunteun
ABSTRAKT
Starch is a major determinant of the glycemic responses elicited by our diets, but the exact contribution of the two main amylolytic enzymes (salivary and pancreatic α-amylases) remains a matter of debate. Our aim was to investigate the contribution of the oral, gastric and intestinal phases to the hydrolysis of starch in bread and pasta during dynamic in vitro digestions using DiDGI®. Before its inactivation by the low gastric pH, salivary α-amylase released about 80% of the starch in bread and 30% of that in pasta, hydrolysing over half of it into oligosaccharides. Accordingly, the contribution of pancreatic α-amylase during the intestinal phase was lower for bread than pasta. Our results are well correlated with in vivo data, and demonstrate the importance of salivary α-amylase during oro-gastric processing of starchy foods. This finding is discussed in relation with observations regarding salivary α-amylase from other fields of knowledge.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications