Merck
  • Identification of N-(5-(phenoxymethyl)-1,3,4-thiadiazol-2-yl)acetamide derivatives as novel protein tyrosine phosphatase epsilon inhibitors exhibiting anti-osteoclastic activity.
Identification of N-(5-(phenoxymethyl)-1,3,4-thiadiazol-2-yl)acetamide derivatives as novel protein tyrosine phosphatase epsilon inhibitors exhibiting anti-osteoclastic activity.
Bioorganic & medicinal chemistry (2018-09-27)
Bonsu Ku, Hye-Yeoung Yun, Kyung Won Lee, Ho-Chul Shin, Sang-Rae Lee, Chang Hyen Kim, Hwangseo Park, Kyu Yang Yi, Chang Hoon Lee, Seung Jun Kim
ABSTRAKT
Cytosolic protein tyrosine phosphatase epsilon (cyt-PTPε) plays a central role in controlling differentiation and function of osteoclasts, whose overactivation causes osteoporosis. Based on our previous study reporting a number of cyt-PTPε inhibitory chemical compounds, we carried out a further and extended analysis of our compounds to examine their effects on cyt-PTPε-mediated dephosphorylation and on osteoclast organization and differentiation. Among five compounds showing target selectivity to cyt-PTPε over three other phosphatases in vitro, two compounds exhibited an inhibitory effect against the dephosphorylation of cellular Src protein, the cyt-PTPε substrate. Moreover, these two compounds caused destabilization of the podosome structure that is necessary for the bone-resorbing activity of osteoclasts, and also attenuated cellular differentiation of monocytes into osteoclasts, without affecting cell viability. Therefore, these findings not only verified anti-osteoclastic effects of our cyt-PTPε inhibitory compounds, but also showed that cyt-PTPε expressed in osteoclasts could be a putative therapeutic target worth considering.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Kit formulated with all liquid reagents
Supelco
Malachite Green chloride, analytical standard