Merck
  • Effects of Interfacial Passivation on the Electrical Performance, Stability, and Contact Properties of Solution Process Based ZnO Thin Film Transistors.
Effects of Interfacial Passivation on the Electrical Performance, Stability, and Contact Properties of Solution Process Based ZnO Thin Film Transistors.
Materials (Basel, Switzerland) (2018-09-21)
Liaojun Wan, Fuchao He, Yu Qin, Zhenhua Lin, Jie Su, Jingjing Chang, Yue Hao
ABSTRAKT
This paper reports low temperature solution processed ZnO thin film transistors (TFTs), and the effects of interfacial passivation of a 4-chlorobenzoic acid (PCBA) layer on device performance. It was found that the ZnO TFTs with PCBA interfacial modification layers exhibited a higher electron mobility of 4.50 cm² V-1 s-1 compared to the pristine ZnO TFTs with a charge carrier mobility of 2.70 cm² V-1 s-1. Moreover, the ZnO TFTs with interfacial modification layers could significantly improve device shelf-life stability and bias stress stability compared to the pristine ZnO TFTs. Most importantly, interfacial modification layers could also decrease the contact potential barrier between the source/drain electrodes and the ZnO films when using high work-function metals such as Ag and Au. These results indicate that high performance TFTs can be obtained with a low temperature solution process with interfacial modification layers, which strongly implies further potential for their applications.