Merck
Polyamines modulate aluminum-induced oxidative stress differently by inducing or reducing H2O2 production in wheat.
Chemosphere (2018-09-03)
Yan Yu, Weiwei Zhou, Kejin Zhou, Wenjing Liu, Xin Liang, Yao Chen, Dasheng Sun, Xianyong Lin
ABSTRAKT
Polyamines are important bioactive molecules involved in regulating H2O2 homeostasis, which is recognized as a major stimulus of oxidative stress under aluminum (Al) exposure. In this study, we investigated the involvement of spermidine oxidation in Al-induced oxidative stress, and its modulation by exogenous putrescine (Put) in two wheat genotypes differing in Al tolerance. Aluminum caused more severe oxidative damage at the root apexes in the Al-sensitive genotype Yangmai-5 than in the tolerant Xi Aimai-1, but these effects were significantly reversed by exogenous Put and polyamine oxidase (PAO) inhibitors. Aluminum caused a more significant increase in cell wall-bound PAO (CW-PAO) activity in Yangmai-5 than in Xi Aimai-1. Inhibiting of CW-PAO reduced H2O2 accumulation, restored Spd decline in both genotypes, indicating its potential role in Al-induced H2O2 production through catalyzing Spd oxidation. Additionally, Al significantly increased the activity of plasma membrane-NADPH oxidase, another H2O2 generator, in wheat roots. Put application significantly inhibited the activity of CW-PAO and plasma membrane-NADPH oxidase, and reduced H2O2 accumulation in Al-stressed wheat roots. Antioxidant enzymes were significantly stimulated by Al, but not Put. Overall, Put may protect wheat roots against Al-induced oxidative stress through regulating H2O2 production by inhibiting CW-PAO and plasma membrane-NADPH oxidase.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium 3,5-dichloro-2-hydroxybenzenesulfonate, used for peroxide measurement
Sigma-Aldrich
1,12-Diaminododecane, 98%
Sigma-Aldrich
Triton X-100, laboratory grade