Merck
  • Contrast-enhanced CT imaging as a non-destructive tool for ex vivo examination of the biochemical content and structure of the human meniscus
Contrast-enhanced CT imaging as a non-destructive tool for ex vivo examination of the biochemical content and structure of the human meniscus
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society
Oh DJ, et al.