Merck
  • Roles of electrostatic interaction and polymer structure in the binding of beta-lactoglobulin to anionic polyelectrolytes: measurement of binding constants by frontal analysis continuous capillary electrophoresis
Roles of electrostatic interaction and polymer structure in the binding of beta-lactoglobulin to anionic polyelectrolytes: measurement of binding constants by frontal analysis continuous capillary electrophoresis
Langmuir (2000)
Hattori T, et al.