Merck
  • Integrin activation or alpha9 expression allows retinal pigmented epithelial cell adhesion on Bruch?s membrane in wet age-related macular degeneration
Integrin activation or alpha9 expression allows retinal pigmented epithelial cell adhesion on Bruch?s membrane in wet age-related macular degeneration
Brain (2010)
Afshari FT, et al.