Merck
  • Insulin activates protein kinases C-zeta and C-lambda by an autophosphorylation-dependent mechanism and stimulates their translocation to GLUT4 vesicles and other membrane fractions in rat adipocytes.
Insulin activates protein kinases C-zeta and C-lambda by an autophosphorylation-dependent mechanism and stimulates their translocation to GLUT4 vesicles and other membrane fractions in rat adipocytes.
The Journal of biological chemistry (1999-08-28)
M L Standaert, G Bandyopadhyay, L Perez, D Price, L Galloway, A Poklepovic, M P Sajan, V Cenni, A Sirri, J Moscat, A Toker, R V Farese
ABSTRAKT
In rat adipocytes, insulin provoked rapid increases in (a) endogenous immunoprecipitable combined protein kinase C (PKC)-zeta/lambda activity in plasma membranes and microsomes and (b) immunoreactive PKC-zeta and PKC-lambda in GLUT4 vesicles. Activity and autophosphorylation of immunoprecipitable epitope-tagged PKC-zeta and PKC-lambda were also increased by insulin in situ and phosphatidylinositol 3,4,5-(PO(4))(3) (PIP(3)) in vitro. Because phosphoinositide-dependent kinase-1 (PDK-1) is required for phosphorylation of activation loops of PKC-zeta and protein kinase B, we compared their activation. Both RO 31-8220 and myristoylated PKC-zeta pseudosubstrate blocked insulin-induced activation and autophosphorylation of PKC-zeta/lambda but did not inhibit PDK-1-dependent (a) protein kinase B phosphorylation/activation or (b) threonine 410 phosphorylation in the activation loop of PKC-zeta. Also, insulin in situ and PIP(3) in vitro activated and stimulated autophosphorylation of a PKC-zeta mutant, in which threonine 410 is replaced by glutamate (but not by an inactivating alanine) and cannot be activated by PDK-1. Surprisingly, insulin activated a truncated PKC-zeta that lacks the regulatory (presumably PIP(3)-binding) domain; this may reflect PIP(3) effects on PDK-1 or transphosphorylation by endogenous full-length PKC-zeta. Our findings suggest that insulin activates both PKC-zeta and PKC-lambda in plasma membranes, microsomes, and GLUT4 vesicles by a mechanism requiring increases in PIP(3), PDK-1-dependent phosphorylation of activation loop sites in PKC-zeta and lambda, and subsequent autophosphorylation and/or transphosphorylation.