Merck
Interleukin-6 directly modulates stem cell factor-dependent development of human mast cells derived from CD34(+) cord blood cells.
Blood (1999-07-09)
T Kinoshita, N Sawai, E Hidaka, T Yamashita, K Koike
ABSTRAKT
In the present study, we attempted to clarify the effects of interleukin-6 (IL-6) on the growth and properties of human mast cells using cultured mast cells selectively generated by stem cell factor (SCF) from CD34(+) cord blood cells. The addition of IL-6 to cultures containing mast cells resulted in a substantial reduction of the number of progenies grown by SCF in the liquid culture. This IL-6-mediated inhibition of mast cell growth may be due in part to the suppression at the precursor level, according to the results of a clonal cell culture assay. Moreover, a flow cytometric analysis showed that the cultured mast cells grown in the presence of SCF+IL-6 had decreased c-kit expression. The exposure of cultured mast cells to SCF+IL-6 also caused substantial increases in the cell size, frequency of chymase-positive cells, and intracellular histamine level compared with the values obtained with SCF alone. The flow cytometric analysis showed low but significant levels of expression of IL-6 receptor (IL-6R) and gp130 on the cultured mast cells grown with SCF. The addition of either anti-IL-6R antibody or anti-gp130 antibody abrogated the biological functions of IL-6. Although IL-4 exerted an effect similar to that of IL-6 on the cultured mast cells under stimulation with SCF, the results of comparative experiments suggest that the two cytokines use different regulatory mechanisms. Taken together, the present findings suggest that IL-6 modulates SCF-dependent human mast cell development directly via an IL-6R-gp130 system.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tryptase Antibody, Mast Cell, clone G3, clone G3, Chemicon®, from mouse