Merck
  • Lipopolysaccharide induces physical proximity between CD14 and toll-like receptor 4 (TLR4) prior to nuclear translocation of NF-kappa B.
Lipopolysaccharide induces physical proximity between CD14 and toll-like receptor 4 (TLR4) prior to nuclear translocation of NF-kappa B.
Journal of immunology (Baltimore, Md. : 1950) (2000-10-18)
Q Jiang, S Akashi, K Miyake, H R Petty
ABSTRAKT
CD14, a GPI-linked protein, plays a pivotal role in LPS-mediated signaling by potentiating leukocyte adherence, activation, and cytokine production. Recent studies have identified the Toll-like receptor 4 (TLR4) as a membrane cofactor in LPS-mediated transmembrane signaling in cytokine induction, although the mechanism responsible for this cooperation is unknown. Using fluorescence resonance energy transfer (RET) techniques, we demonstrate that LPS triggers a physical association between CD14 and TLR4. Because LPS stimulation upregulates CD14 and TLR4 expression, it was necessary to control for the possibility that these newly expressed molecules were associated with one another independent of LPS stimulation. Although the calcium ionophore A23187 increased the expression of CD14 and TLR4, they did not exhibit energy transfer. However, following A23187 treatment, LPS promoted physical proximity between CD14 and TLR4. Therefore, we suggest that a close interaction between CD14 and TLR4 participates in LPS signaling, leading to nuclear translocation of NF-kappaB.