- Cross-coupling of mesylated phenol derivatives with potassium ammonio- and amidomethyltrifluoroborates.
Cross-coupling of mesylated phenol derivatives with potassium ammonio- and amidomethyltrifluoroborates.
Organic letters (2011-02-08)
Gary A Molander, Floriane Beaumard
PMID21294530
ABSTRAKT
A large array of aryl and heteroaryl mesylates have been successfully employed as electrophiles in a Csp(2)-Csp(3) Suzuki-Miyaura cross-coupling with potassium ammonio- and amidomethyltrifluoroborates to afford the corresponding products in high yields.