Merck
Developmental regulation of the NMDA receptor subunits, NR3A and NR1, in human prefrontal cortex.
Cerebral Cortex (1991)
Henson, MA; Roberts, AC; Salimi, K; Vadlamudi, S; Hamer, RM; Gilmore, JH; Jarskog, LF; Philpot, BD
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-NR3A, Upstate®, from rabbit
Sigma-Aldrich
Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse