Merck
Protein 600 is a microtubule/endoplasmic reticulum-associated protein in CNS neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2008-04-04)
Su Yeon Shim, Jian Wang, Naoyuki Asada, Gernot Neumayer, Hong Chi Tran, Kei-ichiro Ishiguro, Kamon Sanada, Yoshihiro Nakatani, Minh Dang Nguyen
ABSTRAKT
There is an increasing body of literature pointing to cytoskeletal proteins as spatial organizers and interactors of organelles. In this study, we identified protein 600 (p600) as a novel microtubule-associated protein (MAP) developmentally regulated in neurons. p600 exhibits the unique feature to interact with the endoplasmic reticulum (ER). Silencing of p600 by RNA interference (RNAi) destabilizes neuronal processes in young primary neurons undergoing neurite extension and containing scarce staining of the ER marker Bip. Furthermore, in utero electroporation of p600 RNAi alters neuronal migration, a process that depends on synergistic actions of microtubule dynamics and ER functions. p600-depleted migrating neurons display thin, crooked, and "zigzag" leading process with very few ER membranes. Thus, p600 constitutes the only known MAP to associate with the ER in neurons, and this interaction may impact on multiple cellular processes ranging from neuronal development to neuronal maturation and plasticity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Endoplasmic Reticulum Isolation Kit, isolation of intact ER from mammalian soft tissues and cultured cells
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®