Merck
An evolutionarily conserved kernel of gata5, gata6, otx2 and prdm1a operates in the formation of endoderm in zebrafish.
Developmental biology (2011-07-16)
Wen-Fang Tseng, Te-Hsuan Jang, Chang-Ben Huang, Chiou-Hwa Yuh
ABSTRAKT
An evolutionarily conserved subcircuit (kernel) dedicated to a specific developmental function is found at the top of the gene regulatory networks (GRNs) hierarchy. Here we comprehensively demonstrate that a pan-deuterostome endoderm specification kernel exists in zebrafish. We analyzed interactions among gata5, gata6, otx2 and prdm1a using specific morpholino knockdowns and measured the gene expression profiles by quantitative real-time RT-PCR and in situ hybridization. The mRNA rescue experiment validated the specificity of the morpholino knockdown. We found that the interactions among gata5, gata6, otx2 and prdm1a determine the initial specification of the zebrafish endoderm. Although otx2 can activate both gata5 and gata6, and the prdm1a/krox homologue also activates some endoderm transcription factors, a feedback loop from Gata to otx2 and prdm1a is missing. Furthermore, we found the positive regulation between gata5 and gata6 to further lock-on the mesendoderm specification by the Gata family. Chromatin immunoprecipitation was used to further validate the recruitment of Otx2 to the gata5 and gata6 loci. Functional assays revealed that module B of gata6 and the basal promoter of gata5 drive the gene at the mesendoderm, and mutational analysis demonstrated that Otx2 and Gata5/6 contribute to reporter gene activation. This is the first direct evidence for evolutionarily conserved endoderm specification across echinoderms and vertebrates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
EZ-Magna ChIP® A - Zestaw do immunoprecypitacji chromatyny, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.
Sigma-Aldrich
EZ-Magna ChIP® G - Zestaw do immunoprecypitacji chromatyny, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic G beads. Control primers included.