Merck
Genetic evidence that brain-derived neurotrophic factor mediates competitive interactions between individual cortical neurons.
Proceedings of the National Academy of Sciences of the USA
English, CN; Vigers, AJ; Jones, KR
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse