Merck
The soybean beta-conglycinin beta 51-63 fragment suppresses appetite by stimulating cholecystokinin release in rats.
The Journal of nutrition (2003-07-31)
Takashi Nishi, Hiroshi Hara, Kozo Asano, Fusao Tomita
ABSTRAKT
We previously demonstrated that soybean beta-conglycinin peptone suppresses food intake and gastric emptying by direct action on rat small intestinal mucosal cells to stimulate cholecystokinin (CCK) release. The aim of the present study was to define the active fragment in beta-conglycinin by using synthetic peptides chosen from the sequence of three beta-conglycinin subunits. We selected the fragments that had multiple nonadjacent arginine residues, and investigated their ability to bind to components of the rat intestinal brush border membrane as well as to stimulate CCK release and appetite suppression. The fragment from 51 to 63 of the beta subunit (beta 51-63) had the strongest binding activity. Intraduodenal infusion of beta 51-63 inhibited food intake and markedly increased portal CCK concentration. The threshold concentration of beta 51-63 to affect food intake was 3 micro mol/L. The CCK-A receptor antagonist abolished the beta 51-63-induced suppression of food intake. Three types of smaller fragments of beta 51-63 (beta 51-59, beta 53-63 and beta 53-59) and two types of fragments similar to beta 51-63 in the beta-conglycinin alpha and alpha' subunits (alpha 212-224 and alpha' 230-240) had less binding ability than did beta 51-63. Model peptides constructed with arginine (R) and glycine (G), such as GRGRGRG, had strong binding affinity, but peptides containing a single R or RR did not. These results indicate that the beta-conglycinin beta 51-63 fragment is the bioactive appetite suppressant in beta-conglycinin, and multiple arginine residues in the fragment may be involved in this effect.