Merck
  • Activation of cellular invasion by trefoil peptides and src is mediated by cyclooxygenase- and thromboxane A2 receptor-dependent signaling pathways.
Activation of cellular invasion by trefoil peptides and src is mediated by cyclooxygenase- and thromboxane A2 receptor-dependent signaling pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2001-06-28)
S Rodrigues, Q D Nguyen, S Faivre, E Bruyneel, L Thim, B Westley, F May, G Flatau, M Mareel, C Gespach, S Emami
ABSTRAKT
We have investigated the possible functional relationships between cellular invasion pathways induced by trefoil factors (TFFs), src, and the cyclooxygenases COX-1 and COX-2. Pharmacological inhibitors of the Rho small GTPase (C3 exoenzyme), phospholipase C (U-73122), cyclooxygenases (SC-560, NS-398), and the thromboxane A2 receptor (TXA2-R) antagonist SQ-295 completely abolished invasion induced by intestinal trefoil factor, pS2, and src in kidney and colonic epithelial cells MDCKts.src and PCmsrc. In contrast, invasion was induced by the TXA2-R mimetic U-46619, constitutively activated forms of the heterotrimeric G-proteins Galphaq (AGalphaq), Galpha12, Galpha13 (AGalpha12/13), which are signaling elements downstream of TXA2-R. Ectopic overexpression of pS2 cDNA and protein in MDCKts.src-pS2 cells and human colorectal cancer cells HCT8/S11-pS2 initiate distinct invasion signals that are Rho independent and COX and TXA2-R dependent. We detected a marked induction of COX-2 protein and accumulation of the stable PGH2/TXA2 metabolite TXB2 in the conditioned medium from cells transformed by src. This led to activation of the TXA2-R-dependent invasion pathway, which is monitored via a Rho- and Galpha12/Galpha13-independent mechanism using the Galphaq/PKC signaling cascade. These findings identify a new intracrine/paracrine loop that can be monitored by TFFs and src in inflammatory diseases and progression of colorectal cancers.