Merck
  • Inhibition of the function of TRPV1-expressing nociceptive sensory neurons by somatostatin 4 receptor agonism: mechanism and therapeutical implications.
Inhibition of the function of TRPV1-expressing nociceptive sensory neurons by somatostatin 4 receptor agonism: mechanism and therapeutical implications.
Current topics in medicinal chemistry (2011-06-16)
János Szolcsányi, Erika Pintér, Zsuzsanna Helyes, Gábor Petho
ABSTRAKT
Release of somatostatin into the circulation from the activated TRPV1-expressing nociceptors revealed by antidromic stimulation of dorsal roots in the rat pinpointed to a novel potential drug target on these nociceptors. The review summarizes the functional, biochemical and pharmacological evidence for a novel somatostatin-mediated counter-regulatory antiinflammatory/antinociceptive "sensocrine" function in rats and guinea-pigs. To identify the somatostatin receptor subtype(s) responsible for this function, experiments were focused on actions of sstR4 receptor agonists as this subtype, similarly to sstR1, is not involved in endocrine regulation. Involvement of somatostatin and the sstR4 was revealed by using pretreatment with somatostatin antibody, depletion of somatostatin with cysteamine, measuring the plasma somatostatin-like immunoreactivity, release from nerves in vitro from isolated trachea, detection of sstR4 receptors in animal and human tissue specimens, using sstR4 gene-deleted mice and investigating in detail effects of a stable peptide analogue of somatostatin (TT-232) and of an ultrapotent non-peptide agonist of sstR4 receptors. Promising antinociceptive, antihyperalgesic effects of these sstR4 agonists were observed in various experimental models of inflammatory and neuropathic conditions which are mediated both by TRPV1-expressing nociceptors and non-neural cells involved in mediation of inflammation. In sstR4 receptor knockout mice an aggravation of inflammation and hyperalgesia was observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cysteamine hydrochloride, BioXtra
Sigma-Aldrich
Cysteamine hydrochloride, ≥97.0% (RT)