Merck
A mutation in the start codon of ?-crystallin D leads to nuclear cataracts in the Dahl SS/Jr-Ctr strain.
Mammalian Genome
Johnson, AC; Lee, JW; Harmon, AC; Morris, Z; Wang, X; Fratkin, J; Rapp, JP; Gomez-Sanchez et al.