Merck
Thermodynamic studies of bovine lung surfactant extract mixing with cholesterol and its palmitate derivative.
Journal of colloid and interface science (2007-04-17)
A K Panda, K Nag, R R Harbottle, F Possmayer, N O Petersen
ABSTRAKT
Langmuir film behavior of bovine lipid extract surfactant (BLES), mixed with cholesterol (CHOL) and cholesterol palmitate (CHOLP), has been studied by surface pressure (pi)-area (A) measurements. Associative interactions, observed for both systems, were less favored at lower BLES content. The presence of unsaturated phospholipids and surfactant proteins in BLES favored the association. Miscibility of BLES was better with CHOLP than with CHOL at all compositions, indicating more compact packing of the BLES-CHOLP than of the BLES-CHOL system. The most stable mixtures were found at 30-40 mol% CHOL and at low pi and at 20-25 mol% CHOLP but at higher pi. These results suggest that BLES-CHOL miscibility is better at low pi and low CHOL concentrations, while BLES-CHOLP miscibility is better at high pi and high CHOLP concentrations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesteryl palmitate, ≥98% (HPLC; detection at 205 nm)