Merck
Retinoblastoma protein functions as a molecular switch determining white versus brown adipocyte differentiation.
Proceedings of the National Academy of Sciences of the USA
Jacob B Hansen, Claus J?rgensen, Rasmus K Petersen, Philip Hallenborg, Rita De Matteis et al.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-Src (Tyr416), klon 9A6, clone 9A6, Upstate®, from mouse