- The induction of collagenase and a neutral proteinase by their high molecular weight substrates in Achromobacter iophagus.
The induction of collagenase and a neutral proteinase by their high molecular weight substrates in Achromobacter iophagus.
Journal of molecular biology (1976-11-05)
V Keil-Dlouha, R Misrahi, B Keil
PMID187752
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Phenylazobenzyloxycarbonyl-Pro-Leu-Gly-Pro-D-Arg trifluoroacetate salt, ≥95% (HPLC)