Merck
  • Identification of glutathione sulfotransferase-pi (GSTP1) as a new resveratrol targeting protein (RTP) and studies of resveratrol-responsive protein changes by resveratrol affinity chromatography.
Identification of glutathione sulfotransferase-pi (GSTP1) as a new resveratrol targeting protein (RTP) and studies of resveratrol-responsive protein changes by resveratrol affinity chromatography.
Anticancer research (2008-04-04)
Tze-Chen Hsieh, Zhirong Wang, Haiteng Deng, Joseph M Wu
ABSTRAKT
Resveratrol shows chemopreventive and other biological affects in in vitro and some animal studies. The bioactivities of resveratrol may be attributed to qualitative and quantitative differences in its cell-type-specific interaction and binding with its cellular targets, denoted as resveratrol targeting proteins (RTPs). To isolate RTPs, resveratrol was linked to epoxy-activated agarose generating an affinity platform to allow the isolation, purification, and characterization of distinct RTPs from cultured prostate cancer cell extracts. Glutathione sulfotransferase-pi (GSTP1) and estrogen receptor-beta (ER-beta) were found to be new RTPs. Resveratrol affinity chromatography was shown to be an easy method for analyzing resveratrol-responsive protein changes in the androgen-dependent LNCaP cells. Resveratrol affects cellular functions at multiple levels, ranging from interaction with detoxification enzymes, such as GSTP1 and transcription by targeting factors such as ER-beta.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Epoxy-activated-Agarose, lyophilized powder
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland