Merck
Dielectric spectroscopy of molecular interactions based on the avidin-biotin complex
Conference proceedings : Annual International Conference of the IEEE Engineering in Medicine and Biology Society (2011)
Mellor, B.L., et al.