Merck
Human immunodeficiency virus type 1 Gag protein binds to cyclophilins A and B.
Cell (1993-06-18)
J Luban, K L Bossolt, E K Franke, G V Kalpana, S P Goff
ABSTRAKT
Retroviral Gag protein is capable of directing the assembly of virion particles independent of other retroviral elements and plays an important role early in the infection of a cell. Using the GAL4 two hybrid system, we screened a cDNA expression library and identified two host proteins, cyclophillins (CyPs) A and B, which interact specifically with the human immunodeficiency virus type 1 (HIV-1) Gag polyprotein Pr55gag. Glutathione S-transferase-CyP fusion proteins bind tightly to Pr55gag in vitro, as well as to the HIV-1 capsid protein p24. Cyclosporin A efficiently disrupts the Gag-CyPA interaction and less efficiently disrupts the Gag-CyPB interaction. The Gag-CyP interaction may be important for the HIV-1 life cycle and may be relevant to the pathology caused by this immunosuppressive virus.