Merck
Properdin plays a protective role in polymicrobial septic peritonitis.
Journal of immunology (Baltimore, Md. : 1950) (2008-02-23)
Cordula M Stover, Jeni C Luckett, Bernd Echtenacher, Aline Dupont, Sue E Figgitt, Jane Brown, Daniela N Männel, Wilhelm J Schwaeble
ABSTRAKT
Properdin is a positive regulator of complement activation so far known to be instrumental in the survival of infections with certain serotypes of Neisseria meningitidis. We have generated a fully backcrossed properdin-deficient mouse line by conventional gene-specific targeting. In vitro, properdin-deficient serum is impaired in alternative pathway-dependent generation of complement fragment C3b when activated by Escherichia coli DH5alpha. Properdin-deficient mice and wild-type littermates compare in their levels of C3 and IgM. In an in vivo model of polymicrobial septic peritonitis induced by sublethal cecal ligation and puncture, properdin-deficient mice appear immunocompromised, because they are significantly impaired in their survival compared with wild-type littermates. We further show that properdin localizes to mast cells and that properdin has the ability to directly associate with E. coli DH5alpha. We conclude that properdin plays a significant role in the outcome of polymicrobial sepsis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Protein G Sepharose, Fast Flow, recombinant, expressed in E. coli, aqueous ethanol suspension
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution