Merck
Carbohydrate binding specificity of recombinant human macrophage beta-glucan receptor dectin-1.
Bioscience, biotechnology, and biochemistry (2009-01-09)
Minoru Ujita, Hiroko Nagayama, Satoko Kanie, Shota Koike, Yoshiko Ikeyama, Takahiro Ozaki, Hiroki Okumura
ABSTRAKT
Human macrophage dectin-1, a type II transmembrane beta-glucan receptor, was expressed as a fusion protein with an N-terminal hexahistidine tag and glutathione S-transferase in an Escherichia coli cell-free translation system, and assayed for binding specificity. Recombinant dectin-1 specifically bound to some beta-glucans, but not to other carbohydrates. The beta-glucan binding of recombinant dectin-1 was inhibited by laminarin, a soluble beta-glucan, and by laminarioligosaccharides, but not by other carbohydrates. These results suggest that recombinant human dectin-1 can be used as a useful probe in identifying ligands in humans and tonic foods due to its strict binding specificity.