Merck
The nuclear orphan receptor Nur77 is a lipotoxicity sensor regulating glucose-induced insulin secretion in pancreatic β-cells.
Molecular endocrinology (Baltimore, Md.) (2012-02-04)
Olivier Briand, Audrey Helleboid-Chapman, Maheul Ploton, Nathalie Hennuyer, Rodolphe Carpentier, François Pattou, Brigitte Vandewalle, Ericka Moerman, Valery Gmyr, Julie Kerr-Conte, Jérôme Eeckhoute, Bart Staels, Philippe Lefebvre
ABSTRAKT
The NR4A orphan nuclear receptors Nur77, Nurr1, and Nor1 exert multiple cellular and metabolic functions. These transcriptional regulators are activated in response to extracellular stresses, including lipotoxic fatty acids (FA) and proinflammatory cytokines. The contribution of NR4As to β-cell pathophysiology is, however, unknown. We have therefore examined the role of NR4As as downstream contributors to FA-induced β-cell dysfunctions. Human pancreatic islets and insulinoma β-cells were used to determine transcriptional programs elicited by NR4A, which were compared to those triggered by palmitate treatment. Functional studies evaluated the consequence of an increased NR4A expression on insulin biosynthesis and secretion and cell viability in insulinoma β-cells. FA and cytokine treatment increased NR4A expression in pancreatic β-cells, with Nur77 being most highly inducible in murine β-cells. Nur77, Nurr1, or Nor1 modulated common and distinct clusters of genes involved notably in cation homeostasis and insulin gene transcription. By altering zinc homeostasis, insulin gene transcription, and secretion, Nur77 was found to be a major transcriptional mediator of part of FA-induced β-cell dysfunctions. The repressive role of Nur77 in insulin gene regulation was tracked down to protein-protein interaction with FoxO1, a pivotal integrator of the insulin gene regulatory network. The present study identifies a member of the NR4A nuclear receptor subclass, Nur77/NR4A1, as a modulator of pancreatic β-cell biology. Together with its previously documented role in liver and muscle, its role in β-cells establishes Nur77 as an important integrator of glucose metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
ChIPAb+ Oczyszczony acetylo-histon H3 (Lys9) - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, from rabbit, purified by using Protein A
Sigma-Aldrich
Sodium palmitate, ≥98.5%