Merck
  • Cultivar discrimination of Portuguese Lupinus albus by seed protein electrophoresis: the importance of considering glutelins and glycoproteins
Cultivar discrimination of Portuguese Lupinus albus by seed protein electrophoresis: the importance of considering glutelins and glycoproteins
Field Crops Research (2004)
Vaz A C, et al.