Merck
Normal human keratinocytes pretreated with damaged mRNA are resistant to UVB damage and exhibit increased DNA repair.
Skin pharmacology and physiology (2004-12-21)
T Mammone, E Goyarts, P E Ya, M Ingrassia, D Gan, K Marenus, D Maes
ABSTRAKT
Treatment of normal human keratinocytes with UVC-irradiated rabbit globin mRNA 24 h before and after UVB exposure increased the survival of the human keratinocytes. We also observed that UVC-damaged mRNA reduced the formation of sunburn cells in skin models. We next tested the effects of UVC-damaged mRNA on cellular repair of DNA. DNA repair was evaluated using 2 assay methods. The first method used a damaged plasmid that is transfected back into the cell where it is repaired by the host cell repair mechanism. In these experiments, we observed that externally added UVC-damaged rabbit globin mRNA enhanced the repair of a plasmid transfected into the host keratinocyte cells. The second method used to determine the effects on DNA repair was direct immunostaining for thymidine-thymidine dimers (TT dimers) in histological sections of the skin models. Skin models were irradiated with UVB and then fixed immediately or after 24 h and stained for TT dimers. UVB irradiation immediately caused an increase in the number of stained keratinocytes in the skin. The number of stained cells decreased in skin fixed 24 h after UVB. This is due to repair of the TT dimers and their removal. Sections of skin models pretreated with UV-damaged mRNA exhibit greater removal of these TT dimers after 24 h. The above evidence suggests that damaged mRNA can trigger a host cell DNA repair pathway.