Merck
  • Base-specific and enantioselective studies for the DNA binding of iron(II) mixed-ligand complexes containing 1,10-phenanthroline and dipyrido[3,2-a:2',3'-c]phenazine.
Base-specific and enantioselective studies for the DNA binding of iron(II) mixed-ligand complexes containing 1,10-phenanthroline and dipyrido[3,2-a:2',3'-c]phenazine.
Spectrochimica Acta Part A: Molecular Spectroscopy (2007)
Mudasir, Wijaya K, Wahyuni ET, et al.