Merck
  • Detection of fetomaternal genotype associations in early-onset disorders: evaluation of different methods and their application to childhood leukemia
Detection of fetomaternal genotype associations in early-onset disorders: evaluation of different methods and their application to childhood leukemia
BioMed Research International (2010)
Healy J, et al.