- In vitro pharmacodynamic characteristics of nystatin including time-kill and postantifungal effect.
In vitro pharmacodynamic characteristics of nystatin including time-kill and postantifungal effect.
Antimicrobial agents and chemotherapy (2000-09-19)
S M Gunderson, H Hoffman, E J Ernst, M A Pfaller, M E Klepser
PMID10991881
ABSTRAKT
Four Candida albicans isolates and six non-albicans Candida isolates were evaluated by time-kill methods to characterize the relationship between nystatin concentrations, the rate and extent of fungicidal activity, and the postantifungal effect (PAFE). Against Candida species, nystatin exhibits concentration-dependent fungicidal activity and a pronounced PAFE.