Merck
  • Fishmeal with different levels of biogenic amines in aquafeed: Comparison of feed protein quality, fish growth performance, and metabolism
Fishmeal with different levels of biogenic amines in aquafeed: Comparison of feed protein quality, fish growth performance, and metabolism
Aquaculture (Amsterdam, Netherlands) (2018)
Jasour M, et al.