Merck
Crystal structures of bovine milk xanthine dehydrogenase and xanthine oxidase: structure-based mechanism of conversion
Proceedings of the National Academy of Sciences of the USA (2000)
Enroth C, et al.