Merck
  • High Mannose Binding Lectin (PFL) from Pseudomonas fluorescens Down-Regulates Cancer-Associated Integrins and Immune Checkpoint Ligand B7-H4.
High Mannose Binding Lectin (PFL) from Pseudomonas fluorescens Down-Regulates Cancer-Associated Integrins and Immune Checkpoint Ligand B7-H4.
Cancers (2019-05-06)
Yuichiro Sato, Kiminori Matsubara, Takanori Kubo, Hirobumi Sunayama, Yuta Hatori, Kinjiro Morimoto, Toshio Seyama
ABSTRAKT
Pseudomonas fluorescens lectin (PFL), which belongs to the high mannose (HM)-binding OAAH (Oscillatoria agardhii agglutinin homologue) lectin family, induces cancer cell death. However, the detailed mechanisms underlying this process have not yet been elucidated. We found that PFL decreased various integrins as well as EGFR in cancer cells by promoting internalization and autophagic degradation of these molecules, subsequently inducing caspase-8 dependent cell apoptosis. As revealed by an ex vivo angiogenesis assay using the rat aortic model, PFL inhibited neovascularization in a dose-dependent manner, which was potentially mediated by down-regulation of endothelium integrins. Interestingly, PFL also down-regulated B7-H4 in cancer cells, which has been implicated as a negative regulator of T cell-mediated immunity. We found that B7-H4 co-localized with β3 integrin in MKN28 gastric cancer cells. siRNA silencing of B7-H4 in MKN28 cells decreased expression of β3 integrin, suggesting physical and functional association between these molecules. Direct interaction of PFL with integrin αvβ3 or B7-H4 was examined by surface plasmon resonance analysis, which detected high affinity glycan-dependent binding to PFL. These investigations suggest that PFL interaction with cell surface integrins is a key process for the anti-cancer activities of PFL.