Merck
  • Protective effects and active ingredients of Salvia miltiorrhiza Bunge extracts on airway responsiveness, inflammation and remodeling in mice with ovalbumin-induced allergic asthma.
Protective effects and active ingredients of Salvia miltiorrhiza Bunge extracts on airway responsiveness, inflammation and remodeling in mice with ovalbumin-induced allergic asthma.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2019-01-03)
Junming Luo, Li Zhang, Xinyi Zhang, Yingying Long, Fang Zou, Chunsong Yan, Wei Zou
ABSTRAKT
Salvia miltiorrhiza Bunge (S. miltiorrhiza), a traditional Chinese medicine, has demonstrated antioxidant, anti-inflammatory, and antibacterial activities. However, its effects against asthma that shows chronic inflammation and oxidative damage remain unknown. To assess the effects of S. miltiorrhiza extracts on airway responsiveness, inflammation, and remodeling in ovalbumin (OVA)-induced asthmatic mice. Mice with ovalbumin (OVA)-induced allergic asthma were treated with S. miltiorrhiza extracts, and airway resistance (RL) to methacholine, inflammatory cell infiltration, Th1/Th2 cytokine levels, and airway remodeling were assessed. TGF-β1-induced BEAS-2B and MRC-5 cells were used to evaluate the effects of five S. miltiorrhiza compounds on epithelial-mesenchymal transition and fibrosis. OVA-challenge resulted in remarkably increased RL, inflammatory cell infiltration, Th1/Th2 cytokine levels in BALF, goblet cell hyperplasia, collagen deposition, and airway wall thickening. Daily treatment with S. miltiorrhiza ethanolic (EE, 246 mg/kg) or water (WE, 156 mg/kg) extract significantly reduced OVA-induced airway inflammatory cell infiltration, Th1/Th2 cytokine amounts, and goblet cells hyperplasia. However, only WE remarkably decreased RL, collagen deposition, and airway wall thickening. Moreover, Chromatography showed that salvianic acid A and caffeic acid levels were much higher in WE than EE, while rosmarinic acid was slightly lower; salvianolic acid B and tanshinone IIA levels were much higher in EE than WE. Interestingly, caffeic acid and rosmarinic acid were more potent in reducing E-cadherin and vimentin levels in TGF-β1-induced BEAS-2B cells, and α-SMA and COL1A1 amounts in TGF-β1-induced MRC-5 cells. Both S. miltiorrhiza WE and EE alleviate airway inflammation in mice with OVA-sensitized allergic asthma. S. miltiorrhiza WE is more potent in reducing responsiveness and airway remodeling.