Merck
Formation, Identification, and Occurrence of New Bromo- and Mixed Halo-Tyrosyl Dipeptides in Chloraminated Water.
Environmental science & technology (2019-02-27)
Guang Huang, Lindsay K Jmaiff Blackstock, Ping Jiang, Zhongshan Liu, Xiufen Lu, Xing-Fang Li
ABSTRAKT
Dipeptides are widely present in surface water and serve as precursors to form disinfection byproducts (DBPs) during disinfection (e.g., chloramination). Bromide (Br-) and iodide (I-) are common in many source waters, enhancing Br- and I-DBP formation. Recently Cl-, I-, and Cl-I-dipeptides were identified after chloramination of tyrosyl dipeptides in the presence of I- and were detected in authentic disinfected drinking water samples. However, the formation and occurrence of Br- and mixed halogen (Cl, Br, and/or I)-dipeptides in disinfected water have not been studied. Here we investigated the formation of halogenated dipeptides from three aromatic dipeptides, phenylalanylglycine (Phe-Gly), tyrosylalanine (Tyr-Ala), and tyrosylglycine (Tyr-Gly), under chloramination in the presence of Br- and I- at environmentally relevant levels ([Br-] and [I-], 0 and 0 μg L-1, 6 and 30 μg L-1, 30 and 30 μg L-1, 150 and 30 μg L-1, 300 and 30 μg L-1, and 900 and 30 μg L-1, respectively). For the first time, N-Br- and N,N-di-Br- as well as N-Br- N-Cl- and N-Br-3-I-tyrosyl dipeptides were identified using infusion electrospray quadrupole time-of-flight mass spectrometry. Tyrosyl dipeptides produced N-Cl-, 3-I-/3,5-di-I-, and N-Cl-3-I-tyrosyl dipeptides, while Phe-Gly formed only N-Cl-/ N, N-di-Cl-Phe-Gly. To determine halogenated dipeptides in authentic water samples, we developed a new method of solid phase extraction and high-performance liquid chromatography with quadrupole ion trap mass spectrometry using reaction monitoring. 3,5-Di-I-Tyr-Ala and N-Br-Tyr-Ala were detected in treated water but not in the corresponding raw water, warranting further investigation into the occurrence of halogenated peptides in other drinking water systems.