Merck
Spiroindolone Analogues as Potential Hypoglycemic with Dual Inhibitory Activity on α-Amylase and α-Glucosidase.
Molecules (Basel, Switzerland) (2019-06-28)
Mezna Saleh Altowyan, Assem Barakat, Abdullah Mohammed Al-Majid, H A Al-Ghulikah
ABSTRAKT
Inhibition of α-amylase and α-glucosidase by specified synthetic compounds during the digestion of starch helps control post-prandial hyperglycemia and could represent a potential therapy for type II diabetes mellitus. A new series of spiroheterocyclic compounds bearing oxindole/benzofuran/pyrrolidine/thiazolidine motifs were synthesized via a 1,3-dipolar cyclo-addition reaction approach. The specific compounds were obtained by reactions of chalcones having a benzo[b]furan scaffold (compounds 2a-f), with a substituted isatin (compounds 3a-c) and heterocyclic amino acids (compounds 4a,b). The target spiroindolone analogues 5a-r were evaluated for their potential inhibitory activities against the enzymes α-amylase and α-glucosidase. Preliminary results indicated that some of the target compounds exhibit promising α-amylase and α-glucosidase inhibitory activity. Among the tested spiroindolone analogues, the cycloadduct 5r was found to be the most active (IC50 = 22.61 ± 0.54 μM and 14.05 ± 1.03 μM) as α-amylase and α-glucosidase inhibitors, with selectivity indexes of 0.62 and 1.60, respectively. Docking studies were carried out to confirm the binding interaction between the enzyme active site and the spiroindolone analogues.