Merck
  • Identification of Two New Phenanthrenes from Dendrobii Herba and Their Cytotoxicity towards Human Hypopharynx Squamous Carcinoma Cell (FaDu).
Identification of Two New Phenanthrenes from Dendrobii Herba and Their Cytotoxicity towards Human Hypopharynx Squamous Carcinoma Cell (FaDu).
Molecules (Basel, Switzerland) (2019-06-28)
Bomi Nam, Seung Mok Ryu, Dongho Lee, Chan-Hun Jung, Chang Hyun Jin, Jin-Baek Kim, Ik-Soo Lee, Ah-Reum Han
ABSTRAKT
Two new phenanthrenes, (1R,2R)-1,7-hydroxy-2,8-methoxy-2,3-dihydrophenanthrene-4(1H)-one (1) and 2,7-dihydroxy-phenanthrene-1,4-dione (2), were isolated from the ethyl acetate-soluble fraction of Dendrobii Herba, together with seven known phenanthrenes (3-9), two bibenzyls (10-12), and a lignan (13). Structures of 1 and 2 were elucidated by analyzing one-dimensional (1D) and two-dimensional (2D)-NMR and High-resolution electrospray ionization mass spectra (HR-ESI-MS) data. The absolute configuration of compound 1 was confirmed by the circular dichroism (CD) spectroscopic method. In cytotoxicity assay using FaDu human hypopharynx squamous carcinoma cell line, compounds 3-6, 8, 10, and 12 showed activities, with IC50 values that ranged from 2.55 to 17.70 μM.