Merck
Mitophagy Failure in APP and Tau Overexpression Model of Alzheimer's Disease.
Journal of Alzheimer's disease : JAD (2019-07-01)
Patricia Martín-Maestro, Ricardo Gargini, Esther García, Diana Simón, Jesús Avila, Vega García-Escudero
ABSTRAKT
Mitochondrial alterations and oxidative stress are common features of Alzheimer's disease brain and peripheral tissues. Moreover, mitochondrial recycling process by autophagy has been found altered in the sporadic form of the disease. However, the contribution of the main proteins involved in this pathology such as amyloid-β protein precursor (AβPP) and tau needs to be achieved. With this aim, human unmodified fibroblasts were transduced with lentivectors encoding APP and Tau and treated with CCCP to study the mitophagy process. Both AβPP and tau separately increased autophagy flux mainly by improving degradation phase. However, in the specific case of mitophagy, labeling of mitochondria by PINK1 and PARK2 to be degraded by autophagy seemed reduced, which correlates with the long-term accumulation of mitochondria. Nevertheless, the combination of tau and AβPP was necessary to cause a mitophagy functional impairment reflected in the accumulation of depolarized mitochondria labeled by PINK1. The overexpression of Tau and APP recapitulates the mitophagy failure previously found in sporadic Alzheimer's disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tau Mouse mAb (TAU-5), liquid, clone TAU-5, Calbiochem®
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-BNIP3 antibody, Mouse monoclonal, clone ANa40, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder