Merck
BCL-2 family protein BOK is a positive regulator of uridine metabolism in mammals.
Proceedings of the National Academy of Sciences of the United States of America (2019-07-18)
Rahul Srivastava, Zhipeng Cao, Christina Nedeva, Samara Naim, Daniel Bachmann, Tatiana Rabachini, Lahiru Gangoda, Sanjay Shahi, Jason Glab, Joseph Menassa, Laura Osellame, Tao Nelson, Yuniel Fernandez-Marrero, Fiona Brown, Andrew Wei, Francine Ke, Lorraine O'Reilly, Marcel Doerflinger, Cody Allison, Andrew Kueh, Rob Ramsay, Brian J Smith, Suresh Mathivanan, Thomas Kaufmann, Hamsa Puthalakath
ABSTRAKT
BCL-2 family proteins regulate the mitochondrial apoptotic pathway. BOK, a multidomain BCL-2 family protein, is generally believed to be an adaptor protein similar to BAK and BAX, regulating the mitochondrial permeability transition during apoptosis. Here we report that BOK is a positive regulator of a key enzyme involved in uridine biosynthesis; namely, uridine monophosphate synthetase (UMPS). Our data suggest that BOK expression enhances UMPS activity, cell proliferation, and chemosensitivity. Genetic deletion of Bok results in chemoresistance to 5-fluorouracil (5-FU) in different cell lines and in mice. Conversely, cancer cells and primary tissues that acquire resistance to 5-FU down-regulate BOK expression. Furthermore, we also provide evidence for a role for BOK in nucleotide metabolism and cell cycle regulation. Our results have implications in developing BOK as a biomarker for 5-FU resistance and have the potential for the development of BOK-mimetics for sensitizing 5-FU-resistant cancers.