Merck
Wet Chemical Method for Black Phosphorus Thinning and Passivation.
ACS applied materials & interfaces (2019-02-12)
Shuangqing Fan, JingSi Qiao, Jiawei Lai, Haicheng Hei, Zhihong Feng, Qiankun Zhang, Daihua Zhang, Sen Wu, Xiaodong Hu, Dong Sun, Wei Ji, Jing Liu
ABSTRAKT
Layered black phosphorus (BP) has been expected to be a promising material for future electronic and optoelectronic applications since its discovery. However, the difficulty in mass fabricating layered air-stable BP severely obstructs its potential industry applications. Here, we report a new BP chemical modification method to implement all-solution-based mass production of layered air-stable BP. This method uses the combination of two electron-deficient reagents 2,2,6,6-tetramethylpiperidinyl- N-oxyl (TEMPO) and triphenylcarbenium tetrafluorobor ([Ph3C]BF4) to accomplish thinning and/or passivation of BP in organic solvent. The field-effect transistor and photodetection devices constructed from the chemically modified BP flakes exhibit enhanced performances with environmental stability up to 4 months. A proof-of-concept BP thin-film transistor fabricated through the all-solution-based exfoliation and modification displays an air-stable and a typical p-type transistor behavior. This all-solution-based method improves the prospects of BP for industry applications.