Merck
Thymoquinone-Induced Tristetraprolin Inhibits Tumor Growth and Metastasis through Destabilization of MUC4 mRNA.
International journal of molecular sciences (2019-05-31)
Se-Ra Lee, Jeong-Yeon Mun, Mi-So Jeong, Hyun-Hee Lee, Yun-Gil Roh, Won-Tae Kim, Min-Hye Kim, Jeonghoon Heo, Yung Hyun Choi, Su Jin Kim, Hee-Jae Cha, Mira Jun, Sun-Hee Leem
ABSTRAKT
Tristetraprolin (TTP), a well-characterized AU-rich element (ARE) binding protein, functions as a tumor suppressor gene. The purpose of this study was to investigate whether a bioactive substance derived from a natural medicinal plant affects the induction of TTP and to elucidate its mechanism. We examined the effects of natural bioactive materials including Resveratrol (RSV), thymoquinone (TQ) and curcumin on the expression of TTP in cancer cell. TQ derived from a natural plant Nigella sativa increased the expression levels of TTP mRNA and proteins in a dose-dependent manner in gastric and breast cancer cells. TQ-induced TTP increased the instability of MUC4 mRNA by direct binding of TTP to ARE in the 3'UTR of MUC4 mRNA. The induction of TTP by TQ also reduced the proliferation, migration and invasion of cancer cells. The expression of the epithelial-mesenchymal (EMT)-related genes, which were target genes of TTP, was also decreased by the TQ treatment. In the in vivo experiments using mouse melanoma cells, TQ-induced TTP inhibited metastasis of tumor cells. We have found that TQ-induced TTP might inhibit metastasis by reducing tumor cell migration and invasion through destabilization of MUC4 mRNA, which suggest the MUC4 as a novel target to TTP.