Merck
  • The role of UCH-L1, MMP-9, and GFAP as peripheral markers of different susceptibility to seizure development in a preclinical model of epilepsy.
The role of UCH-L1, MMP-9, and GFAP as peripheral markers of different susceptibility to seizure development in a preclinical model of epilepsy.
Journal of neuroimmunology (2019-04-06)
Natalia Chmielewska, Piotr Maciejak, Danuta Turzyńska, Alicja Sobolewska, Aleksandra Wisłowska-Stanek, Karolina Kołosowska, Adam Płaźnik, Janusz Szyndler
ABSTRAKT
In our study, we assessed the potency of the brain-derived proteins ubiquitin carboxy-terminal hydrolase L1 (UCH-L1), matrix metalloproteinase 9 (MMP-9), glial fibrillary acidic protein (GFAP) and the immune activation indicators interleukin 1β (IL-1β) and interleukin 6 (IL-6) as peripheral biomarkers of different susceptibilities to kindling in a preclinical model. We observed increased plasma UCH-L1 levels in kindled vs. control animals. Furthermore, MMP-9 and IL-1β concentrations were the lowest in rats resistant to kindling. In summary, UCH-L1 is an indicator of neuronal loss and BBB disruption after seizure. MMP-9 and IL-1β may indicate resistance to kindling. UCH-L1, MMP-9 and IL-1β may have utility as peripheral biomarkers with translational potency in the clinic.