Merck
Absorption, distribution, metabolism, and excretion of mTOR kinase inhibitor CC-223 in rats, dogs, and humans.
Xenobiotica; the fate of foreign compounds in biological systems (2017-12-06)
Zeen Tong, Christian Atsriku, Usha Yerramilli, Xiaomin Wang, Jim Nissel, Yan Li, Sekhar Surapaneni
ABSTRAKT
1. The absorption, distribution, metabolism, and excretion of CC-223 were studied following a single oral dose of [14C]CC-223 to rats (3 mg/kg; 90 μCi/kg), dogs (1.5 mg/kg; 10 μCi/kg), and healthy volunteers (20 mg; 200 nCi). 2. CC-223-derived radioactivity was widely distributed in rats. Excretion of radioactivity was rapid and nearly complete from rats (87%), dogs (78%), and humans (97%). Feces was the major excretion pathway for rats (67%) and dogs (70%), whereas urine (57.6%) was the major elimination route for humans. Urine and bile each contained approximately 20% administered radioactivity in rats, whereas bile (20%) played a more important role than urine (<10%) in the excretion of absorbed radioactivity in dogs. Based on excretion data, CC-223 had good absorption, with greater than 56%, 29%, and 57% of the oral dose absorbed in rats, dogs, and humans, respectively. 3. CC-223 was the prominent radioactive component in circulation of rats (>71% of the exposure to total radioactivity) and dogs (≥45.5%), whereas M1 (76.5%) was the predominant circulating metabolite in humans. M1 and M1-derived metabolites accounted for >66% of human dose. CC-223 was extensively metabolized in rats, dogs, and humans through glucuronidation, O-demethylation, oxidation, and combinations of these pathways.