Merck
Effects of aflatoxin B1 on the cell cycle distribution of splenocytes in chickens.
Journal of toxicologic pathology (2019-02-12)
Hang Li, Ke Guan, Zhicai Zuo, Fengyuan Wang, Xi Peng, Jing Fang, Hengmin Cui, Yi Zhou, Ping Ouyang, Gang Su, Zhengli Chen
ABSTRAKT
The purpose of the present study was to evaluate effects of aflatoxin B1 (AFB1) on the cell cycle and proliferation of splenic cells in chickens. A total of 144 one-day-old Cobb male chickens were randomly divided into 2 equal groups of 72 each and were fed on diets as follows: a control diet and a 0.6 mg/kg AFB1 diet for 21 days. The AFB1 diet reduced body weight, absolute weight and relative weight of the spleen in broilers. Histopathological lesions in AFB1 groups were characterized as slight congestion in red pulp and lymphocytic depletion in white pulp. Compared with the control group, the expression levels of ataxia-telangiectasia mutated (ATM), cyclin E1, cyclin-dependent kinases 6 (CDK6), CDK2, p53, p21 and cyclin B3 mRNA were significantly increased, while the mRNA expression levels of cyclin D1, cdc2 (CDK1), p16, p15 were significantly decreased in the AFB1 groups. Significantly decreased proliferating cell nuclear antigen (PCNA) expression and arrested G0G1 phases of the cell cycle were also seen in the AFB1 groups. In conclusion, dietary AFB1 could induce cell cycle blockage at G0G1 phase and impair the immune function of the spleen. Cyclin D1/CDK6 complex, which inhibits the activin/nodal signaling pathway, might play a significant role in the cell cycle arrest induced by AFB1.